siRNA Details
| Gene Symbol: Acer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290541.1 | NP_001277470.1 | 1 | 174 | ||
| NM_001290543.1 | NP_001277472.1 | 1 | 174 | |||
| NM_139306.3 | NP_647467.1 | 1 | 174 | |||
| GenBank mRNA | AF282864.1 | - | 1 | 133 | ||
| AK085306.1 | - | 1 | 170 | |||
| AY312515.1 | - | 1 | 99 | |||
| BC059819.1 | - | 1 | 174 | 6839525 | ||
| CB723138.1 | - | 1 | 174 | |||

Acer2
Genomic Map