siRNA Details
| Gene Symbol: Leprot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175036.4 | NP_778201.1 | 4 | 553 | ||
| GenBank mRNA | AJ011565.1 | - | 4 | 526 | ||
| AK009569.1 | - | 4 | 554 | |||
| AK088848.1 | - | 2 | 370 | |||
| AK152008.1 | - | 4 | 619 | |||
| AK159489.1 | - | 4 | 619 | |||
| AK160453.1 | - | 4 | 567 | |||
| AK165821.1 | - | 4 | 612 | |||
| AK168597.1 | - | 4 | 589 | |||
| BC004744.1 | - | 4 | 564 | 3583760 | ||
| BC010289.1 | - | 4 | 474 | 2651399 | ||

Leprot
Genomic Map