siRNA Details
| Gene Symbol: C8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290645.1 | NP_001277574.1 | 7 | 1116 | ||
| NM_146148.2 | NP_666260.1 | 8 | 1213 | |||
| GenBank mRNA | AB077295.1 | - | 8 | 1227 | ||
| AB077296.1 | - | 8 | 1176 | |||
| AB077297.1 | - | 7 | 1082 | |||
| AB077298.1 | - | 8 | 1176 | |||
| AK090245.1 | - | 8 | 1213 | |||
| BC027748.1 | - | 8 | 1209 | 5052412 | ||

C8a
Genomic Map