siRNA Details
| Gene Symbol: Ttc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177667.4 | NP_808335.1 | 5, 6 | 1130 | ||
| GenBank mRNA | AK028895.1 | - | 5, 6 | 1131 | ||
| BC096484.1 | - | 5, 6 | 1107 | |||
| BC132162.1 | - | 5, 6 | 1071 | 40130439 | ||

Ttc22
Genomic Map