siRNA Details
| Gene Symbol: Cyb5rl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175471.2 | NP_780680.1 | 2 | 469 | ||
| GenBank mRNA | AK053031.1 | - | 5 | 873 | ||
| AK220154.1 | - | 3 | 371 | |||
| AY437136.1 | - | 299 | ||||
| BC004750.1 | - | 2 | 308 | |||
| BC043687.1 | - | 2 | 469 | 5136046 | ||

Cyb5rl
Genomic Map