siRNA Details
Gene Symbol: Cyb5rl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175471.2 | NP_780680.1 | 4 | 708 | ||
GenBank mRNA | AK042316.1 | - | 5 | 2323 | ||
AK053031.1 | - | 7 | 1113 | |||
AK220154.1 | - | 5 | 610 | |||
BC043687.1 | - | 4 | 708 | 5136046 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
