siRNA Details
| Gene Symbol: Yipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205156.1 | NP_001192085.1 | 4 | 420 | ||
| NM_145550.3 | NP_663525.1 | 5 | 501 | |||
| GenBank mRNA | AK081205.1 | - | 5 | 488 | ||
| BC009080.1 | - | 5 | 433 | 2655774 | ||

Yipf1
Genomic Map