siRNA Details
| Gene Symbol: Glis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147221.2 | NP_671754.2 | 5 | 1556 | ||
| GenBank mRNA | AF220434.1 | - | 3 | 1410 | ||
| AF486579.1 | - | 5 | 1555 | |||
| AK145342.1 | - | 2 | 80 | |||
| BC066157.1 | - | 3 | 991 | 30468539 | ||
| JQ043365.1 | - | 4 | 1335 | |||

Glis1
Genomic Map