siRNA Details
| Gene Symbol: Zcchc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175472.3 | NP_780681.2 | 23 | 4124 | ||
| GenBank mRNA | AK077768.1 | - | 1 | 3558 | ||
| AK134388.1 | - | 10 | 1380 | |||
| AK220327.1 | - | 21 | 3372 | |||
| BC145507.1 | - | 21 | 3705 | |||
| BC150791.1 | - | 22 | 3775 | 9087702 | ||

Zcchc11
Genomic Map