siRNA Details
| Gene Symbol: Ttc39a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145948.1 | NP_001139420.1 | 15 | 1357 | ||
| NM_153392.2 | NP_700441.2 | 15 | 1366 | |||
| GenBank mRNA | AK048659.1 | - | 15 | 1367 | ||
| AK076641.1 | - | 15 | 1320 | |||
| AK076720.1 | - | 15 | 1359 | |||
| AK076772.1 | - | 15 | 1321 | |||
| AK172951.1 | - | 15 | 1289 | |||
| BC023883.1 | - | 15 | 1365 | 5150841 | ||
| BC026660.1 | - | 15 | 1307 | 4988900 | ||

Ttc39a
Genomic Map