siRNA Details
| Gene Symbol: Slc5a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145551.4 | NP_663526.3 | 4, 5 | 462 | ||
| GenBank mRNA | AK029158.1 | - | 4, 5 | 463 | ||
| AK033425.1 | - | 3, 4 | 392 | |||
| AK050696.1 | - | 3, 4 | 342 | |||
| BC021357.1 | - | 4, 5 | 443 | 5012356 | ||

Slc5a9
Genomic Map