siRNA Details
| Gene Symbol: Slc5a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145551.4 | NP_663526.3 | 5 | 554 | ||
| GenBank mRNA | AK029158.1 | - | 5 | 555 | ||
| AK033425.1 | - | 4 | 484 | |||
| AK050696.1 | - | 4 | 434 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a9| Gene Symbol: Slc5a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145551.4 | NP_663526.3 | 5 | 554 | ||
| GenBank mRNA | AK029158.1 | - | 5 | 555 | ||
| AK033425.1 | - | 4 | 484 | |||
| AK050696.1 | - | 4 | 434 | |||