siRNA Details
| Gene Symbol: Cyp4a29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100183.1 | NP_001093653.1 | 12 | 1482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4a29| Gene Symbol: Cyp4a29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100183.1 | NP_001093653.1 | 12 | 1482 | ||