siRNA Details
| Gene Symbol: PLGLB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002665.4 | NP_002656.1 | 4 | 1264 | ||
| GenBank mRNA | AF075095.1 | - | 1 | 146 | ||
| Gene Symbol: PLGLB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001032392.2 | NP_001027564.1 | 4 | 1260 | ||

PLGLB2
Genomic Map