siRNA Details
| Gene Symbol: Kdm4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161823.1 | NP_001155295.1 | 19 | 3023 | ||
| NM_172382.2 | NP_759014.2 | 19 | 3023 | |||
| GenBank mRNA | AK054095.1 | - | 19 | 2987 | ||
| AK129187.1 | - | 19 | 2878 | |||
| AK136085.1 | - | 10 | 1680 | |||
| AK145066.1 | - | 19 | 3009 | |||
| AK145947.1 | - | 19 | 2945 | |||
| BC028866.1 | - | 19 | 2926 | 4207552 | ||

Kdm4a
Genomic Map