siRNA Details
| Gene Symbol: AU022252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012400.2 | NP_001012400.1 | 3 | 283 | ||
| GenBank mRNA | AK158349.1 | - | 3 | 241 | ||
| AK167445.1 | - | 3 | 245 | |||
| BC035522.1 | - | 3 | 266 | |||
| BC089614.1 | - | 3 | 265 | 6748514 | ||

AU022252
Genomic Map