siRNA Details
| Gene Symbol: Rhbdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183163.2 | NP_898986.2 | 5 | 684 | ||
| GenBank mRNA | BX511550.1 | - | 4 | 462 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhbdl2| Gene Symbol: Rhbdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183163.2 | NP_898986.2 | 5 | 684 | ||
| GenBank mRNA | BX511550.1 | - | 4 | 462 | ||