siRNA Details
| Gene Symbol: Rhbdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183163.2 | NP_898986.2 | 8 | 958 | ||
| GenBank mRNA | BC052650.1 | - | 8 | 1372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhbdl2| Gene Symbol: Rhbdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183163.2 | NP_898986.2 | 8 | 958 | ||
| GenBank mRNA | BC052650.1 | - | 8 | 1372 | ||