siRNA Details
| Gene Symbol: Oscp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172701.2 | NP_766289.2 | 4, 5 | 639 | ||
| GenBank mRNA | AB154826.1 | - | 4, 5 | 592 | ||
| AB286960.1 | - | 5, 6 | 640 | |||
| AK077467.1 | - | 4, 5 | 605 | |||
| AK077909.1 | - | 4, 5 | 560 | |||
| BC045150.1 | - | 4, 5 | 585 | 6309923 | ||

Oscp1
Genomic Map