siRNA Details
| Gene Symbol: Zfp362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081098.1 | NP_001074567.1 | 3 | 247 | ||
| GenBank mRNA | BC147222.1 | - | 2 | 146 | 9056279 | |
| BC147223.1 | - | 2 | 146 | 9056281 | ||
| BC150964.1 | - | 2 | 146 | 9087875 | ||

Zfp362
Genomic Map