siRNA Details
| Gene Symbol: Adgrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199696.1 | NP_001186625.1 | 13 | 2648 | ||
| NM_001290714.1 | NP_001277643.1 | 13 | 2856 | |||
| NM_001290715.1 | NP_001277644.1 | 13 | 2583 | |||
| NM_173071.3 | NP_775094.2 | 14 | 2813 | |||
| GenBank mRNA | AK144268.1 | - | 5 | 698 | ||
| AK144559.1 | - | 5 | 698 | |||
| AK147485.1 | - | 13 | 2857 | |||
| AK158735.1 | - | 13 | 2459 | |||
| AY168407.1 | - | 13 | 2566 | |||
| BC056926.1 | - | 14 | 2727 | 5705854 | ||
More Information
Set Membership: |
|

Adgrb2
Genomic Map