siRNA Details
| Gene Symbol: Fam76a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163792.1 | NP_001157264.1 | 4 | 453 | ||
| NM_145553.2 | NP_663528.1 | 4 | 453 | |||
| GenBank mRNA | AK037828.1 | - | 1 | 138 | ||
| AK038831.1 | - | 4 | 461 | |||
| AK154575.1 | - | 4 | 454 | |||
| BC008163.1 | - | 4 | 413 | 3709256 | ||

Fam76a
Genomic Map