siRNA Details
| Gene Symbol: Ahdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146155.3 | NP_666267.3 | 6 | 4245 | ||
| GenBank mRNA | AK083440.1 | - | 1 | 1506 | ||
| AK134372.1 | - | 1 | 1799 | |||
| BC042621.1 | - | 6 | 4234 | 4911638 | ||
| BC060231.1 | - | 5 | 4213 | 6852595 | ||

Ahdc1
Genomic Map