siRNA Details
| Gene Symbol: Ahdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146155.3 | NP_666267.3 | 6 | 1348 | ||
| GenBank mRNA | AK149347.1 | - | 6 | 1477 | ||
| BC042621.1 | - | 6 | 1334 | 4911638 | ||
| BC060231.1 | - | 5 | 1316 | 6852595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahdc1| Gene Symbol: Ahdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146155.3 | NP_666267.3 | 6 | 1348 | ||
| GenBank mRNA | AK149347.1 | - | 6 | 1477 | ||
| BC042621.1 | - | 6 | 1334 | 4911638 | ||
| BC060231.1 | - | 5 | 1316 | 6852595 | ||