siRNA Details
| Gene Symbol: Pigv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145955.1 | NP_001139427.1 | 3 | 485 | ||
| NM_178698.5 | NP_848813.3 | 3 | 1331 | |||
| GenBank mRNA | AK046527.1 | - | 3 | 1292 | ||
| AK047322.1 | - | 3 | 1129 | |||
| AK052446.1 | - | 3 | 1305 | |||
| AK080858.1 | - | 3 | 1285 | |||
| BC055060.1 | - | 3 | 1281 | 6528864 | ||

Pigv
Genomic Map