siRNA Details
| Gene Symbol: Aim1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162970.1 | NP_001156442.1 | 16 | 4049 | ||
| GenBank mRNA | AK145690.1 | - | 16 | 4055 | ||
| AK155034.1 | - | 4 | 400 | |||
| AK165037.1 | - | 3 | 324 | |||
| BC042709.1 | - | 11 | 1070 | |||
| BC150738.1 | - | 15 | 3978 | 9087649 | ||

Aim1l
Genomic Map