siRNA Details
| Gene Symbol: Grhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013756.1 | NP_001013778.1 | 9 | 1323 | ||
| GenBank mRNA | BC089372.1 | - | 9 | 1323 | 30607679 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grhl3| Gene Symbol: Grhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013756.1 | NP_001013778.1 | 9 | 1323 | ||
| GenBank mRNA | BC089372.1 | - | 9 | 1323 | 30607679 | |