siRNA Details
| Gene Symbol: Emc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039200.2 | NP_001034289.1 | 11 | 1165 | ||
| NM_146157.4 | NP_666269.2 | 11 | 1165 | |||
| GenBank mRNA | AK049061.1 | - | 11 | 1156 | ||
| AK077496.1 | - | 11 | 1156 | |||
| AK089289.1 | - | 11 | 1144 | |||
| AK129052.1 | - | 11 | 1118 | |||
| AK138637.1 | - | 11 | 1138 | |||
| AK161677.1 | - | 11 | 1128 | |||
| AK170739.1 | - | 11 | 1144 | |||
| BC056952.1 | - | 6 | 613 | |||
| BC129804.1 | - | 11 | 1152 | 40087024 | ||
| BC139366.1 | - | 11 | 1151 | 8862388 | ||

Emc1
Genomic Map