siRNA Details
| Gene Symbol: Emc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039200.2 | NP_001034289.1 | 23 | 2859 | ||
| NM_146157.4 | NP_666269.2 | 24 | 2868 | |||
| GenBank mRNA | AF525925.1 | - | 14 | 1707 | ||
| AK049061.1 | - | 24 | 2859 | |||
| AK077496.1 | - | 24 | 2859 | |||
| AK129052.1 | - | 24 | 2821 | |||
| AK161677.1 | - | 23 | 2824 | |||
| BC056952.1 | - | 19 | 2588 | |||
| BC129804.1 | - | 24 | 2855 | 40087024 | ||
| BC139366.1 | - | 24 | 2854 | 8862388 | ||

Emc1
Genomic Map