siRNA Details
| Gene Symbol: Crocc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145958.1 | NP_001139430.1 | 15 | 2043 | ||
| NM_172122.2 | NP_742120.2 | 16 | 2296 | |||
| GenBank mRNA | AF527975.1 | - | 16 | 2296 | ||
| AK122285.1 | - | 8 | 1046 | |||
| BC036729.1 | - | 1 | 34 | |||
| BC054054.1 | - | 14 | 2024 | 6394697 | ||

Crocc
Genomic Map