siRNA Details
| Gene Symbol: Fbxo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176848.1 | NP_789818.1 | 6 | 1073 | ||
| GenBank mRNA | AK140175.1 | - | 6 | 1117 | ||
| BC027053.1 | - | 5 | 1026 | |||
| BC046586.1 | - | 6 | 1073 | 6487759 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo2| Gene Symbol: Fbxo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176848.1 | NP_789818.1 | 6 | 1073 | ||
| GenBank mRNA | AK140175.1 | - | 6 | 1117 | ||
| BC027053.1 | - | 5 | 1026 | |||
| BC046586.1 | - | 6 | 1073 | 6487759 | ||