siRNA Details
| Gene Symbol: PLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002667.4 | NP_002658.1 | 2 | 301 | ||
| GenBank mRNA | AK311799.1 | - | 2 | 236 | ||
| BC005269.1 | - | 2 | 236 | 3934199 | ||
| BP257094.1 | - | 2 | 301 | |||
| BP259965.1 | - | 2 | 285 | |||
| M60411.1 | - | 1 | 148 | |||
| M63603.1 | - | 2 | 255 | |||

PLN
Genomic Map