siRNA Details
| Gene Symbol: Dnajc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172704.3 | NP_766292.2 | 1 | 81 | ||
| GenBank mRNA | AK053156.1 | - | 1 | 63 | ||
| AK089983.1 | - | 1 | 73 | |||
| AK156793.1 | - | 1 | 63 | |||
| AK171781.1 | - | 1 | 62 | |||
| AK172542.1 | - | 1 | 62 | |||
| BC075619.1 | - | 1 | 57 | |||
| BC079852.1 | - | 1 | 51 | |||
| BC085257.1 | - | 1 | 55 | 5414963 | ||
| BY784439.1 | - | 1 | 91 | |||

Dnajc11
Genomic Map