siRNA Details
| Gene Symbol: Cep104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177673.2 | NP_808341.1 | 7 | 780 | ||
| GenBank mRNA | AK080736.1 | - | 1 | 70 | ||
| AK129163.1 | - | 6 | 646 | |||
| AK143802.1 | - | 7 | 1195 | |||
| AK149826.1 | - | 7 | 1131 | |||
| BC046331.1 | - | 7 | 780 | 6485859 | ||

Cep104
Genomic Map