siRNA Details
| Gene Symbol: Plekhn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008233.3 | NP_001008234.2 | 13 | 1438 | ||
| GenBank mRNA | AK048084.1 | - | 11 | 3417 | ||
| BC025458.1 | - | 3 | 312 | |||
| BC046544.1 | - | 13 | 1438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhn1| Gene Symbol: Plekhn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008233.3 | NP_001008234.2 | 13 | 1438 | ||
| GenBank mRNA | AK048084.1 | - | 11 | 3417 | ||
| BC025458.1 | - | 3 | 312 | |||
| BC046544.1 | - | 13 | 1438 | |||