siRNA Details
| Gene Symbol: Klhl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198305.2 | NP_938047.1 | 7 | 1171 | ||
| GenBank mRNA | AK049214.1 | - | 7 | 1048 | ||
| AK154523.1 | - | 7 | 1109 | |||
| AK170825.1 | - | 7 | 1109 | |||
| AK194598.1 | - | 2 | 86 | |||
| AY423764.1 | - | 7 | 1149 | |||
| BC139309.1 | - | 7 | 1112 | 8862331 | ||

Klhl17
Genomic Map