siRNA Details
| Gene Symbol: Insig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153526.5 | NP_705746.1 | 2 | 401 | ||
| GenBank mRNA | AF527630.1 | - | 2 | 415 | ||
| AK088380.1 | - | 2 | 448 | |||
| AK135941.1 | - | 2 | 452 | |||
| AK135989.1 | - | 2 | 454 | |||
| AK139913.1 | - | 2 | 451 | |||
| AK151186.1 | - | 2 | 451 | |||
| BC079559.1 | - | 2 | 401 | |||
| BC132167.1 | - | 2 | 409 | 40130444 | ||
| BC132499.1 | - | 2 | 409 | 40130776 | ||

Insig1
Genomic Map