siRNA Details
| Gene Symbol: Hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289798.1 | NP_001276727.1 | 14 | 1348 | ||
| NM_001289799.1 | NP_001276728.1 | 14 | 1344 | |||
| NM_145558.2 | NP_663533.1 | 14 | 1301 | |||
| GenBank mRNA | AK033462.1 | - | 14 | 1283 | ||
| AK076814.1 | - | 14 | 1264 | |||
| AK083164.1 | - | 14 | 1309 | |||
| AK083767.1 | - | 14 | 1264 | |||
| AK150889.1 | - | 14 | 1263 | |||
| AK168594.1 | - | 13 | 1128 | |||
| AK169637.1 | - | 14 | 1277 | |||
| BC005585.1 | - | 13 | 1232 | 3158015 | ||

Hadhb
Genomic Map