siRNA Details
| Gene Symbol: Agbl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048192.2 | NP_001041657.1 | 10 | 1869 | ||
| NM_001294262.1 | NP_001281191.1 | 9 | 1897 | |||
| NM_174849.3 | NP_777274.1 | 9 | 1786 | |||
| GenBank mRNA | AK029676.1 | - | 9 | 1787 | ||
| AK034894.1 | - | 9 | 1875 | |||
| AK035680.1 | - | 7 | 1354 | |||
| AK083656.1 | - | 8 | 2040 | |||
| BC057349.1 | - | 9 | 1787 | 6813901 | ||
| DQ867034.1 | - | 9 | 1782 | |||
| DQ867035.1 | - | 9 | 1782 | |||
| DQ867036.1 | - | 10 | 1869 | |||
| DQ867037.1 | - | 9 | 1782 | |||
| DQ867038.1 | - | 9 | 1782 | |||

Agbl5
Genomic Map