siRNA Details
| Gene Symbol: Fam193a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243123.1 | NP_001230052.1 | 15 | 2936 | ||
| GenBank mRNA | BC037112.1 | - | 12 | 2073 | ||
| BC099925.1 | - | 12 | 1973 | 30653351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam193a| Gene Symbol: Fam193a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243123.1 | NP_001230052.1 | 15 | 2936 | ||
| GenBank mRNA | BC037112.1 | - | 12 | 2073 | ||
| BC099925.1 | - | 12 | 1973 | 30653351 | ||