siRNA Details
| Gene Symbol: Cytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081106.1 | NP_001074575.1 | 3 | 339 | ||
| GenBank mRNA | AK076770.1 | - | 2 | 231 | ||
| BC063103.1 | - | 3 | 299 | |||
| BC116224.1 | - | 3 | 313 | 40091368 | ||
| BC116225.1 | - | 3 | 313 | 40091369 | ||
| EF108311.1 | - | 3 | 310 | |||

Cytl1
Genomic Map