siRNA Details
| Gene Symbol: Tapt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173764.3 | NP_776125.2 | 9 | 1030 | ||
| GenBank mRNA | AK030002.1 | - | 4 | 272 | ||
| BC096038.1 | - | 6 | 509 | |||
| BC110670.1 | - | 8 | 751 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tapt1| Gene Symbol: Tapt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173764.3 | NP_776125.2 | 9 | 1030 | ||
| GenBank mRNA | AK030002.1 | - | 4 | 272 | ||
| BC096038.1 | - | 6 | 509 | |||
| BC110670.1 | - | 8 | 751 | |||