siRNA Details
| Gene Symbol: Sel1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172710.3 | NP_766298.2 | 9 | 1615 | ||
| GenBank mRNA | AK030487.1 | - | 9 | 1480 | ||
| AK037220.1 | - | 9 | 1479 | |||
| AK122362.1 | - | 9 | 1615 | |||
| BC094607.1 | - | 8 | 1160 | 6837048 | ||
| BC117915.1 | - | 8 | 1116 | 40097358 | ||
| BC117916.1 | - | 8 | 1118 | 40097359 | ||

Sel1l3
Genomic Map