siRNA Details
| Gene Symbol: Atp10d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153389.3 | NP_700438.3 | 2 | 490 | ||
| GenBank mRNA | AJ441079.1 | - | 2 | 490 | ||
| AK036646.1 | - | 1 | 155 | |||
| AK087333.1 | - | 2 | 506 | |||
| BC099527.1 | - | 2 | 480 | |||
| BC141328.1 | - | 2 | 341 | 9055940 | ||

Atp10d
Genomic Map