siRNA Details
| Gene Symbol: Aasdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173765.3 | NP_776126.1 | 4 | 588 | ||
| GenBank mRNA | AB095954.1 | - | 4 | 605 | ||
| AK038779.1 | - | 5 | 598 | |||
| AK043807.1 | - | 4 | 566 | |||
| AK136849.1 | - | 4 | 586 | |||
| AK148405.1 | - | 4 | 587 | |||
| BC058102.1 | - | 3 | 500 | |||
| BC094507.1 | - | 3 | 365 | |||
| BC128330.1 | - | 3 | 460 | 40110405 | ||

Aasdh
Genomic Map