siRNA Details
| Gene Symbol: Tmprss11d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145561.2 | NP_663536.1 | 8, 9 | 993 | ||
| GenBank mRNA | AB053953.1 | - | 8, 9 | 1027 | ||
| AF448809.1 | - | 8, 9 | 972 | |||
| AF539752.1 | - | 3, 4 | 566 | |||
| BC020151.1 | - | 8, 9 | 993 | 4021767 | ||

Tmprss11d
Genomic Map