siRNA Details
| Gene Symbol: PLRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201564.1 | NP_001188493.1 | 10, 11 | 1040 | ||
| NM_002669.3 | NP_002660.1 | 10, 11 | 1067 | |||
| GenBank mRNA | AF044333.1 | - | 9, 10 | 942 | ||
| AK002108.1 | - | 10, 11 | 1042 | |||
| AK307997.1 | - | 9, 10 | 816 | |||
| BC020786.1 | - | 10, 11 | 1004 | 4809482 | ||
| BC106004.1 | - | 10, 11 | 1005 | 3889109 | ||

PLRG1
Genomic Map