siRNA Details
| Gene Symbol: Ythdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177680.3 | NP_808348.2 | 3, 4 | 543 | ||
| GenBank mRNA | AK135309.1 | - | 3, 4 | 799 | ||
| BC044650.1 | - | 2, 3 | 415 | |||
| CA317046.1 | - | 2, 3 | 355 | |||
| CN528111.1 | - | 3, 4 | 465 | |||

Ythdc1
Genomic Map