siRNA Details
| Gene Symbol: PLRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201564.1 | NP_001188493.1 | 5 | 491 | ||
| NM_002669.3 | NP_002660.1 | 5 | 518 | |||
| GenBank mRNA | AF044333.1 | - | 4 | 393 | ||
| AK002108.1 | - | 5 | 493 | |||
| AK307997.1 | - | 5 | 465 | |||
| BC020786.1 | - | 5 | 455 | 4809482 | ||
| BC106004.1 | - | 5 | 456 | 3889109 | ||

PLRG1
Genomic Map