siRNA Details
| Gene Symbol: Parm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145562.2 | NP_663537.1 | 2 | 429 | ||
| GenBank mRNA | AK078862.1 | - | 2 | 391 | ||
| AK078943.1 | - | 2 | 443 | |||
| AK148300.1 | - | 2 | 477 | |||
| AK161172.1 | - | 392 | ||||
| AK167402.1 | - | 2 | 418 | |||
| BC006604.1 | - | 2 | 429 | 3966312 | ||
| EF564364.1 | - | 2 | 214 | |||

Parm1
Genomic Map